CAPTURE BEAD MOUSE GM-CSF A10 13X 70UL
Catalog # CA77759-730
Supplier: Biolegend
New Product
CAPTURE BEAD MOUSE GM-CSF A10 13X 70UL
Catalog # CA77759-730
Supplier: Biolegend
Supplier Number: 741353
New Product
Frequently Bought Together
Barium chloride dihydrate ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Single-Use Needles, BD Medical
Supplier: BD
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
Acetone ≥99.5% ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Trisodium phosphate dodecahydrate 98.0-102.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Supplier: Corning
First Aid Central Federal Regulation First Aid Kits, Acme United
Supplier: ACME UNITED LIMITED CA
Reagent Sets for Colorimeters, Hach
Supplier: Hach
MinION™ Mk1B DNA/RNA Sequencing Device, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
LabKlenz™ 250 Acid Detergents
Supplier: Steris
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics