CAPTURE BEAD LPLEX HU IL-5 A10 13X 70UL
Catalog # CA77759-804
Supplier: Biolegend
New Product
CAPTURE BEAD LPLEX HU IL-5 A10 13X 70UL
Catalog # CA77759-804
Supplier: Biolegend
Supplier Number: 741406
New Product
Frequently Bought Together
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Sodium sulfate, anhydrous ≥99.0%, granular, Tracepur® 10-60 mesh, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Single-Use Needles, BD Medical
Supplier: BD
Tru-Punch™ Sterile Disposable Biopsy Punches, Sklar
Supplier: Sklar
Autoclave Wraps, HPK Industries
Supplier: HPK Industries
Formaldehyde 10% (v/v) in aqueous solution histological fixative, (non-buffered)
Supplier: Ricca Chemical
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health