LPLEX SCD40L TNFSF5 CAP BDS A7 13X 70UL
Catalog # CA77759-676
Supplier: Biolegend
New Product
LPLEX SCD40L TNFSF5 CAP BDS A7 13X 70UL
Catalog # CA77759-676
Supplier: Biolegend
Supplier Number: 741315
New Product
Frequently Bought Together
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Methanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
Glycerine ≥99.7% ACS, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
COD Digestion vials, VWR Chemicals BDH®
Supplier: VWR International
Pierce™ Blocker™ Casein in PBS, Thermo Scientific
Supplier: Thermo Scientific
Antibiotics and Antimycotics, BD Diagnostics
Supplier: BD
Indicator Tapes, Sklar
Supplier: Sklar
AimTab™ Ketone Tablets, Germaine® Laboratories
Supplier: Germaine Laboratories
Sol-Guard® Safety TB Syringes, Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Sodium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC