CAPTURE BEAD PDGF-BB B5 13X 70UL
Catalog # CA77759-890
Supplier: Biolegend
New Product
CAPTURE BEAD PDGF-BB B5 13X 70UL
Catalog # CA77759-890
Supplier: Biolegend
Supplier Number: 741464
New Product
Frequently Bought Together
Leakbuster™ Specimen Containers
Supplier: Starplex
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Supplier: AMX INDUSTRIAL TRADE MX
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
Carbon Steel Surgical Blades, Non Sterile, Cincinnati Surigcal
Supplier: CINCINNATI SURGICAL CO.
Anti-CD81 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 5A6]
Supplier: Biolegend
Cotton Tip Applicators, DUKAL™ Corporation
Supplier: Dukal
QUICK-LOAD® Taq 2X Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
Adlerkreutz Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
Magnesium
Supplier: Ward's Science
Sodium chloride ≥99.5%, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International
SWAB CLEANER KIT PK50
Supplier: Advanced Instruments
Sodium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC