CAPTURE BEAD HU IFN-A2 B4 13X 70UL
Catalog # CA77759-654
Supplier: Biolegend
New Product
CAPTURE BEAD HU IFN-A2 B4 13X 70UL
Catalog # CA77759-654
Supplier: Biolegend
Supplier Number: 741277
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Single-Use Needles, BD Medical
Supplier: BD
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
Sternheimer Malbin urine sediment stain
Supplier: Globe Scientific
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma