CAPTURE BEAD HU IFN-B B6 13X 70UL
Catalog # CA77759-656
Supplier: Biolegend
New Product
CAPTURE BEAD HU IFN-B B6 13X 70UL
Catalog # CA77759-656
Supplier: Biolegend
Supplier Number: 741278
New Product
Frequently Bought Together
Nitric acid 68 - 70% ACS
Supplier: Anachemia
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
VWR® Disposable Petri Dishes
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Sandwich Bags, Low Density Flip Top Elkay Plastics
Supplier: Elkay Plastics
Hexacarbonylmolybdenum 98%
Supplier: Thermo Scientific Chemicals
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics
Ionic strength adjustor (ISA) solution for use with sodium ion selective electrode
Supplier: Thermo Scientific Chemicals