ANTIBODY 538 MU IGG1 Κ ITCL 100TEST
Catalog # CA77756-778
Supplier: Biolegend
New Product
ANTIBODY 538 MU IGG1 Κ ITCL 100TEST
Catalog # CA77756-778
Supplier: Biolegend
Supplier Number: 402418
New Product
Frequently Bought Together
VWR® Razor Blades
Supplier: VWR International
Peroxide Test, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Single-Use Needles, BD Medical
Supplier: BD
Acetone ACS
Supplier: INNOVA CHEMICALS CA
Cryoscope Standards, Advanced Instruments
Supplier: Advanced Instruments
Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CL17 DPD Indicator, Hach
Supplier: Hach
Maxtaform Square Mesh Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
BUFFER SOLN TOT CHLORINE 473ML
Supplier: Hach
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC