CAPTURE BEAD HU GM-CSF B2 13X 70UL
Catalog # CA77759-806
Supplier: Biolegend
New Product
CAPTURE BEAD HU GM-CSF B2 13X 70UL
Catalog # CA77759-806
Supplier: Biolegend
Supplier Number: 741407
New Product
Frequently Bought Together
Medicine Cups, DUKAL™ Corporation
Supplier: Dukal
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Supplier: MilliporeSigma
Calcium chloride, anhydrous ≥96% ACS
Supplier: VWR
Disposable Biopsy Punches, Integra™ Miltex®
Supplier: Miltex
Masterflex® I/P® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
Water, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
HENKE-JECT® 3-Part Disposable Syringes, Air-Tite
Supplier: Air-Tite
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Supplier: TCI America
tri-Sodium citrate dihydrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Toluene ACS
Supplier: INNOVA CHEMICALS CA
Acetic acid glacial 99.5-100.5% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Blunt Needle Luer Stub Adaptors, SAI Infusion Technologies
Supplier: SAI Infusion Technologies