ANTIBODY 570 AR HAMSTER IGG ITCL 100UG
CA77756-700
New Product
Frequently Bought Together
Masterflex® L/S® Rigid PTFE-Tubing Pump Head, Avantor®
: Avantor Fluid Handling
Hydrochloric acid 36,5 - 38,0% ACS
: Anachemia
Sodium hydroxide 0.100 N (N/10) in aqueous solution
: Ricca Chemical
Ammonium sulfate ≥99.0% ACS, VWR Chemicals BDH®
: VWR International
VWR® Multistage Gas Regulators with Neoprene Diaphragms
: VWR International
Potassium dihydrogen phosphate ≥99.0% ACS, VWR Chemicals BDH®
: VWR International
B352 Specimen Bottles and Caps, Simport Scientific
: Simport Scientific
MICROLITER® 700 Series Syringes, Hamilton
: Hamilton
VWR® PBS (Phosphate Buffered Saline) Buffer Powder Packs
: VWR International
VWR® Premium Dissecting Scissors with Sharp/Blunt Blades
: VWR International
DNA ScreenTape Analysis
: AGILENT TECHNOLOGIES (GENOMICS) CA
PEI MAX® Transfection Reagents (Linear Polyethylenimine Hydrochloride)
: KYFORA BIO LLC.
Human Recombinant CNTF (from <i>E. coli</i>)
: Peprotech
Histosette® I Tissue Processing/Embedding Cassettes, Simport
: Simport Scientific
Water Agar, HiMedia Laboratories
: HiMedia