CAPTURE BEAD LPX NHP TNF-A A6 13X 70UL
Catalog # CA77759-834
Supplier: Biolegend
New Product
CAPTURE BEAD LPX NHP TNF-A A6 13X 70UL
Catalog # CA77759-834
Supplier: Biolegend
Supplier Number: 741421
New Product
Frequently Bought Together
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Sodium molybdate(VI) dihydrate 99.5-103.0% ACS
Supplier: Thermo Scientific Chemicals
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Immunoreagents
Micrewtube® Microcentrifuge Tube with Screw Cap, Simport Scientific
Supplier: Simport Scientific
Tissue Marking Dyes, Azer Scientific
Supplier: Azer Scientific
Pierce™ Blocker™ Casein in PBS, Thermo Scientific
Supplier: Thermo Scientific
Slide Tray Racks and Aluminum Slide Trays, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Adlerkreutz Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
Toluene ACS
Supplier: INNOVA CHEMICALS CA
tert-Butyl methyl ether ≥99% ACS, VWR Chemicals BDH®
Supplier: VWR International
Water, OmniPur® nuclease-free, sterile, DEPC treated, Sigma-Aldrich®
Supplier: MilliporeSigma