CAPTURE BEAD CXCL8 IL-8 CB B7 13X 70UL
Catalog # CA77759-588
Supplier: Biolegend
New Product
CAPTURE BEAD CXCL8 IL-8 CB B7 13X 70UL
Catalog # CA77759-588
Supplier: Biolegend
Supplier Number: 741240
New Product
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
Leakbuster™ Specimen Containers
Supplier: Starplex
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® Specimen Containers, PP with PE Caps
Supplier: VWR International
MICROLITER® 700 Series Syringes, Hamilton
Supplier: Hamilton
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
BPMO ≥99% (by 1H-NMR), high purity
Supplier: Enzo Life Sciences
Saline, 0.85%
Supplier: Hardy Diagnostics
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
VWR® Reagent alcohol
Supplier: VWR International
Ward's® Live<i> E. coli </i>cultures for Coliphage Use
Supplier: Avantor