ANTIBODY MOUSE G-CSF BEAD B5 13X 70UL
Catalog # CA77759-736
Supplier: Biolegend
New Product
ANTIBODY MOUSE G-CSF BEAD B5 13X 70UL
Catalog # CA77759-736
Supplier: Biolegend
Supplier Number: 741356
New Product
Frequently Bought Together
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
TETRAHYDROFURAN OMNISOLV 4L
Supplier: MilliporeSigma
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Convertors® Paper/Film Gas/Steam Sterilization Pouches, Cardinal Health®
Supplier: Cardinal Health
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Vinyl Powder-Free Disposable Gloves, Tronex®
Supplier: Tronex
L(-)-Proline 99%
Supplier: Thermo Scientific Chemicals
Formaldehyde 10% (v/v) in aqueous solution histological fixative, (non-buffered)
Supplier: Ricca Chemical
BUFFER SOLN TOT CHLORINE 473ML
Supplier: Hach
Chloroform ≥99.8% stabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma
tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC