ANTIBODY B1473 MU IGG2B Κ ITCL 10UG
Catalog # CA77756-930
Supplier: Biolegend
New Product
ANTIBODY B1473 MU IGG2B Κ ITCL 10UG
Catalog # CA77756-930
Supplier: Biolegend
Supplier Number: 402719
New Product
Frequently Bought Together
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
Accessories for Osmometer, Advanced Instruments
Supplier: Advanced Instruments
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Single-Use Needles, BD Medical
Supplier: BD
Human Recombinant CNTF (from <i>E. coli</i>)
Supplier: Peprotech
STORAGE SOLUTION PH ELECTRODE
Supplier: Oakton
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics
VWR® Blotting Paper, Grade 703
Supplier: VWR International
Trimming Blades, Avantik
Supplier: Avantik
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
COMPLEMENT GUINEA PIG NON-STERILE 10ML
Supplier: Rockland Immunochemical