LPX MU IL-4 CAPTURE BEAD B5 13X 70UL
Catalog # CA77759-316
Supplier: Biolegend
New Product
LPX MU IL-4 CAPTURE BEAD B5 13X 70UL
Catalog # CA77759-316
Supplier: Biolegend
Supplier Number: 741020
New Product
Frequently Bought Together
VWR® Self-Seal Sterilization Pouches
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
VWR® Specimen Containers with Dual Click
Supplier: VWR International
Leakbuster™ Specimen Containers
Supplier: Starplex
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
VWR® Reagent alcohol
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Miltex® BiopBlade, Integra
Supplier: Miltex
Glutaric acid ≥99.0% (by GC, titration analysis)
Supplier: TCI America
Horse Red Blood Cells 10% Washed Pooled Cells, Rockland Immunochemicals
Supplier: Rockland Immunochemical
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Corning® Cell Recovery Solution, Corning
Supplier: Corning