CAPTURE BEAD HU IL-6 A4 13X 70UL
Catalog # CA77759-622
Supplier: Biolegend
New Product
CAPTURE BEAD HU IL-6 A4 13X 70UL
Catalog # CA77759-622
Supplier: Biolegend
Supplier Number: 741261
New Product
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
Single-Use Needles, BD Medical
Supplier: BD
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Leakbuster™ Specimen Containers
Supplier: Starplex
Peroxide Test, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
Methanol ≥99.8% ACS for HPLC, Supelco®
Supplier: MilliporeSigma
Polysorbate 20, Reagent Grade
Supplier: VWR
Prinzing® Ball Valve Lockouts, Brady Worldwide®
Supplier: Brady Worldwide
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
Coomassie® brilliant blue R-250
Supplier: TCI America
Paraformaldehyde 16% (w/v) in aqueous solution methanol-free
Supplier: Thermo Scientific Chemicals