ANTIBODY MOUSE IFN CPT BD A4 13X 70UL
Catalog # CA77759-636
Supplier: Biolegend
New Product
ANTIBODY MOUSE IFN CPT BD A4 13X 70UL
Catalog # CA77759-636
Supplier: Biolegend
Supplier Number: 741268
New Product
Frequently Bought Together
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® ClikSeal Specimen Containers
Supplier: VWR International
Difco™ Tryptic Soy Agar, BD Biosciences
Supplier: BD
Disposable Biopsy Punches, Integra™ Miltex®
Supplier: Miltex
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
Tissue Marking Dyes (TMD™), TBS®, General Data
Supplier: General Data Healthcare
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
D-(+)-Trehalose dihydrate ≥99.0%, Reagent Grade
Supplier: VWR
Gastight® 1700 Series Syringes, Hamilton
Supplier: Hamilton
TruLine pH 15 Electrode, YSI
Supplier: YSI
Aniline blue 2.5% in 2% acetic acid
Supplier: Electron Microscopy Sciences
Acetic acid glacial 99.5-100.5% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Ammonium iron (II) sulfate hexahydrate 99%
Supplier: Thermo Scientific Chemicals
Sulfuric acid 0.035 N volumetric solution
Supplier: Hach
Phenolphthalein
Supplier: Ward's Science