CAPTURE BEAD CCL4 MIP-1B B9 13X 70UL
Catalog # CA77759-298
Supplier: Biolegend
New Product
CAPTURE BEAD CCL4 MIP-1B B9 13X 70UL
Catalog # CA77759-298
Supplier: Biolegend
Supplier Number: 740998
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Cotton Tip Applicators, DUKAL™ Corporation
Supplier: Dukal
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Cytology Funnels for Shandon CytoSpin™ Centrifuge, Biomedical Polymers
Supplier: Biomedical Polymers
Methanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
Gauze Pads, DUKAL™ Corporation
Supplier: Dukal
VWR® ClikSeal Specimen Containers
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
Sodium chloride, VWR® USP, Ph. Eur., BP, JP endotoxin tested
Supplier: Avantor Performance Materials
Acetone ≥99.5% ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Bulk Syringes, Nonsterile, BD Medical
Supplier: BD
Flanged Plug Caps, Tiered, 13 mm, Globe Scientific
Supplier: Globe Scientific
Reagent Sets for Colorimeters, Hach
Supplier: Hach
Phosphate Buffered Saline (PBS), Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite