LPX MU CXCL10 CAPTURE BEAD A10 13X 70UL
Catalog # CA77759-310
Supplier: Biolegend
New Product
LPX MU CXCL10 CAPTURE BEAD A10 13X 70UL
Catalog # CA77759-310
Supplier: Biolegend
Supplier Number: 741017
New Product
Frequently Bought Together
Waste Gas Control Supplies, VetEquip®
Supplier: Vetequip
Sterile IPA 70% Spray, ITW Texwipe
Supplier: ITW Texwipe
VWR® Disposable Petri Dishes
Supplier: VWR International
Toluene ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
Masterflex® Transfer Tubing, C-Flex®, Clear, Avantor®
Supplier: VWR International
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Supplier: Starplex
MICROLITER® 700 Series Syringes, Hamilton
Supplier: Hamilton
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Orion™ Sure-Flow® pH Electrode 9172BNWP, Thermo Scientific
Supplier: Thermo Fisher Scientific
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: HARDY DIAGNOSTICS CA
NORM-JECT® Luer Lock Bulk Non Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
<i>E. coli</i> Exonuclease I
Supplier: New England Biolabs (NEB)
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
DPD Total Chlorine Reagent Powder Pillows, 5 ml, Hach
Supplier: Hach
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning
E.Z.N.A.® Plasmid Midi Kit, Omega Bio-tek®
Supplier: Omega Bio-Tek