ANTIBODY 570 ANTI-B-AMYLOID 17-24 25UG
Catalog # CA77760-520
Supplier: Biolegend
New Product
ANTIBODY 570 ANTI-B-AMYLOID 17-24 25UG
Catalog # CA77760-520
Supplier: Biolegend
Supplier Number: 800721
New Product
Frequently Bought Together
VWR® Razor Blades
Supplier: VWR International
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Supplier: Thermo Scientific
Leakbuster™ Specimen Containers
Supplier: Starplex
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Single-Use Needles, BD Medical
Supplier: BD
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Circular Cover Glass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences