ANTIBODY GMP RECOM HU FN-1 CF 100UG
Catalog # CA77760-054
Supplier: Biolegend
New Product
ANTIBODY GMP RECOM HU FN-1 CF 100UG
Catalog # CA77760-054
Supplier: Biolegend
Supplier Number: 775316
New Product
Frequently Bought Together
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
VWR® Razor Blades
Supplier: VWR International
Glucose Tolerance Drinks, Azer Scientific
Supplier: Azer Scientific
Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®
Supplier: VWR International
Bulk Syringes, Nonsterile, BD Medical
Supplier: BD
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Single-Use Needles, BD Medical
Supplier: BD
Phenols, CHEMets Visual Kit, CHEMetrics
Supplier: CHEMetrics
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
XLD ( Xylose Lysine Deoxycholate) Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
VWR® Premium Dissecting Scissors with Sharp/Blunt Blades
Supplier: VWR International
Ammonium thiocyanate standardised solution 0.1 N in aqueous solution
Supplier: Thermo Scientific Chemicals
Technical Buffer Solutions, Sachets
Supplier: Mettler Toledo
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International