GMP RECOM HU FGF-BASIC 146 AA CF 50UG
CA77760-358
New Product
Frequently Bought Together
Leakbuster™ Specimen Containers
: Starplex
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
Water, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
: VWR International
Acetic acid glacial ≥99.7% for ICP, for trace metal analysis, J.T.Baker®
: AVANTOR PERFORMANCE MATERIAL LLC
Phenolphthalein 1% (w/v) in 95% (v/v) alcohol indicator, neutralized
: Ricca Chemical
B352 Specimen Bottles and Caps, Simport Scientific
: Simport Scientific
VWR® Premium Dissecting Scissors with Sharp/Blunt Blades
: VWR International
ACK Lysing Buffer, Quality Biological
: QUALITY BIOLOGICAL, INC.
Circular Cover Glass, Electron Microscopy Sciences
: Electron Microscopy Sciences
Acetone ACS
: INNOVA CHEMICALS CA
1,1'-Ferrocenedicarboxylic acid 98%
: Thermo Scientific Chemicals
GASTIGHT® 1000 Series Syringes, Hamilton
: Hamilton