CV GMP RECOM HU FGF-4 CARRIER-FREE 50UG
Catalog # CA77757-948
Supplier: Biolegend
New Product
CV GMP RECOM HU FGF-4 CARRIER-FREE 50UG
Catalog # CA77757-948
Supplier: Biolegend
Supplier Number: 592224
New Product
Frequently Bought Together
SYRINGE STERILE WITHOUT NEEDLE 10ML
Supplier: AMX INDUSTRIAL TRADE MX
Pocket Colorimeter II Test Kits
Supplier: Hach
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Dimethyl sulfoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: VWR International
N,N-Dimethylformamide ≥99.8% (by GC), GR ACS, Supelco®
Supplier: MilliporeSigma
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Immunoreagents
Silica Gel–Desiccant, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Sodium chloride ≥99.5%, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Glutaraldehyde 25% in aqueous solution, EM Grade
Supplier: Electron Microscopy Sciences
Sodium chloride 32.5% (w/v) saturated aqueous solution
Supplier: Ricca Chemical
Diethyl phthalate ≥99.0%
Supplier: TCI America
Anti-TH Mouse Monoclonal Antibody [clone: 2/40/15]
Supplier: Biolegend