CV GMP RECOM HU IL-27 CARRIER-FREE 100UG
Catalog # CA77757-942
Supplier: Biolegend
New Product
CV GMP RECOM HU IL-27 CARRIER-FREE 100UG
Catalog # CA77757-942
Supplier: Biolegend
Supplier Number: 589216
New Product
Frequently Bought Together
VWR® PCR Workstations
Supplier: VWR International
Leakbuster™ Specimen Containers
Supplier: Starplex
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Glucose Tolerance Drinks, Azer Scientific
Supplier: Azer Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
VWR® Reagent alcohol
Supplier: VWR International
HyClone™ SFM4CHO media, Cytiva
Supplier: Cytiva
1,4-Dioxane 99+% ACS
Supplier: Thermo Scientific Chemicals
Gastight® 1800 Series Syringes, Hamilton
Supplier: Hamilton
VWR® General Purpose Dissecting Scissors
Supplier: VWR International
Formaldehyde 10% (v/v) in aqueous solution histological fixative, (non-buffered)
Supplier: Ricca Chemical
Puritan® MALDI-TOF Pointed Applicator Stick, Puritan Medical Products
Supplier: Puritan Medical Products
HistoGel™ Specimen Processing Gel, Epredia
Supplier: Thermo Scientific
Cardboard Freezer Boxes and Dividers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Hydrochloric acid 3.00 N
Supplier: Ricca Chemical
TapeStation Systems Parts and Accessories
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA