MOJOSORT HU CD81 EXOSOME ISOL KIT KT1
Catalog # 77757-440
Supplier: Biolegend
New Product
MOJOSORT HU CD81 EXOSOME ISOL KIT KT1
Catalog # 77757-440
Supplier: Biolegend
Supplier Number: 480174
New Product
Frequently Bought Together
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
BD Microtainer® Tubes, BD Diagnostics
Supplier: BD
Leakbuster™ Specimen Containers
Supplier: Starplex
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Diisopropyl azodicarboxylate 94%
Supplier: Thermo Scientific Chemicals
VACUUBRAND® ME 1C Oil-Free Chemistry Diaphragm Pump, Trade-Up Promotion
Supplier: Brandtech
Diethyl phthalate ≥99.0%
Supplier: TCI America
EMS Swiss Line Standard Tweezer Style 2A, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
Puritan® MALDI-TOF Pointed Applicator Stick, Puritan Medical Products
Supplier: Puritan Medical Products
Silica Gel–Desiccant, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Circular Cover Glass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Quinoline 98%
Supplier: Thermo Scientific Chemicals