SPKYG 570 RAT IGG2B Κ ISOTYPE CTRL 25UG
Catalog # 77756-932
Supplier: Biolegend
New Product
SPKYG 570 RAT IGG2B Κ ISOTYPE CTRL 25UG
Catalog # 77756-932
Supplier: Biolegend
Supplier Number: 402803
New Product
Frequently Bought Together
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Leakbuster™ Specimen Containers
Supplier: Starplex
1X PBS (Phosphate Buffered Saline), pH 7.4
Supplier: GROWCELLS
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
TETRAHYDROFURAN OMNISOLV 4L
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
DNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning
L(-)-Glutathione (reduced form) 97%
Supplier: Thermo Scientific Chemicals
Sodium hydroxide standardised solution 0.1 N in aqueous solution
Supplier: Thermo Scientific Chemicals
Corning® Cell Recovery Solution, Corning
Supplier: Corning
Petroleum spirit, 35…60 °C ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC