ANTIBODY PE F744 RAT IGG2A Κ ITCL 100UG
Catalog # 77756-900
Supplier: Biolegend
New Product
ANTIBODY PE F744 RAT IGG2A Κ ITCL 100UG
Catalog # 77756-900
Supplier: Biolegend
Supplier Number: 402626
New Product
Frequently Bought Together
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Leakbuster™ Specimen Containers
Supplier: Starplex
Alcotabs® Critical Cleaning Detergent Tablets
Supplier: Alconox
Phenolphthalein 1% in alcohol analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
SYRINGE STERILE WITHOUT NEEDLE 10ML
Supplier: AMX INDUSTRIAL TRADE MX
Methanol ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
VWR® Multistage Gas Regulators with Stainless Steel Diaphragms
Supplier: VWR International
Precision pH Test Strips
Supplier: Precision Laboratories
GelRed® & GelGreen® Nucleic Acid Gel Stains, Biotium
Supplier: Biotium
Buffers, Alkaline, Reference Standards, RICCA Chemical Company
Supplier: Ricca Chemical
Sodium chloride ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Pharmacy Bags, Reclosable, Elkay Plastics
Supplier: Elkay Plastics
ES 7X™ Concentrate, MP Biomedicals
Supplier: MP Biomedicals
Budget Antibiotic Disks
Supplier: Ward's Science
Square Petri Dish, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Gastight® 1700 Series Syringes, Hamilton
Supplier: Hamilton