ANTIBODY SPARK YG 570 MU IGG2A ITCL 25UG
Catalog # 77756-830
Supplier: Biolegend
New Product
ANTIBODY SPARK YG 570 MU IGG2A ITCL 25UG
Catalog # 77756-830
Supplier: Biolegend
Supplier Number: 402515
New Product
Frequently Bought Together
Quality Economy Brand Luer Slip Syringes, Air-Tite
Supplier: Air-Tite
Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
2-Propanol ≥99.8% (by GC), HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
VACUUM PUMP VACUUBRAND ME1C
Supplier: Brandtech
Tissue Marking Dyes, Azer Scientific
Supplier: Azer Scientific
Magnesium chloride, anhydrous 99%
Supplier: Thermo Scientific Chemicals
Sodium Bicarbonate
Supplier: Ward's Science
Cetrimide Select Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Technical Buffer Solutions, Sachets
Supplier: Mettler Toledo
InLab® Conductivity Probes, Mettler Toledo
Supplier: Mettler Toledo
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris
Oakton® pH 450 Waterproof Portable Meter, Antylia Scientific
Supplier: Antyila Scientific
Chloroform ≥99.8% (chloroform + ethanol basis) stabilized, GR ACS, Supelco®
Supplier: MilliporeSigma
RedDot™1 Far-Red Nuclear Stain, 200X in Water, Biotium
Supplier: Biotium