ANTIBODY SPKYG 570 RAT IGG2A ITCL 100UG
77756-872
New Product
Frequently Bought Together
INCYTO C-Chip™ Disposable Hemacytometers, SKC
: MEDONE SURGICAL, INC
DIBMA Screening Kit, HEPES
: CUBE BIOTECH
High Sensitivity RNA ScreenTape Analysis
: AGILENT TECHNOLOGIES (GENOMICS) CA
Hydrochloric acid 3.00 N NF, USP
: Ricca Chemical
Puritan® MALDI-TOF Pointed Applicator Stick, Puritan Medical Products
: Puritan Medical Products
2,2'-Bipyridyl 99+%
: Thermo Scientific Chemicals
Disposable Base Molds, Simport Scientific
: Simport Scientific
Ethylene glycol ≥99% ACS, Laboratory Reagent, VWR Chemicals BDH®
: VWR International
Ascorbic Acid
: Ward's Science
STORAGE SOLUTION PH ELECTRODE
: Oakton
Norcamphor 98+%
: Thermo Scientific Chemicals
Dirhenium heptaoxide ≥99.9% (metals basis)
: Thermo Scientific Chemicals
Parafilm M, Electron Microscopy Sciences
: Electron Microscopy Sciences