ANTIBODY 570 AR HAMSTER IGG ITCL 25UG
Catalog # 77756-698
Supplier: Biolegend
New Product
ANTIBODY 570 AR HAMSTER IGG ITCL 25UG
Catalog # 77756-698
Supplier: Biolegend
Supplier Number: 400981
New Product
Frequently Bought Together
Hydrion™, Mikro pH Test Paper with Dispensor, Micro Essential Laboratory
Supplier: Micro Essential
Nitric acid 68 - 70% ACS
Supplier: Anachemia
Cotton Tip Applicators, DUKAL™ Corporation
Supplier: Dukal
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
LaMotte® InstaTest® Water Test Strips
Supplier: LaMotte
VWR® Specimen Containers with Dual Click
Supplier: VWR International
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
Hydrochloric acid standardised solution 0.5 N
Supplier: Thermo Scientific Chemicals
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: HARDY DIAGNOSTICS CA
SILICA GEL IRREGULAR SILIAFLASH P60 25KG
Supplier: SiliCycle
Bromothymol blue 0.04% (w/v) in aqueous solution indicator
Supplier: Ricca Chemical
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA