Abgene™ Sample Processing and Storage Plates
Product Details & Documents
Abgene™ polypropylene MicroWell and DeepWell plates provide performance and value for sample storage, high-throughput screening, and molecular diagnostic assays. Abgene™ plates are made with high-quality, medical-grade, virgin polypropylene resin. These plates benefit from exceptional design and manufacturing advancements that ensure sample integrity and facilitate workflow efficiency.
Made of virgin, medical-grade polypropylene
- hDNA-free, DNase-/RNase-free, and PCR inhibitor-free
- Low extractables and leachables
- 48 to 384 well plate formats available
- Temperature range: −80 to +121 °C
- Volume range: 55 µl to 6 ml
- ANSI/SLAS-compliant
Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries, or storing samples for either intermediate or long-term use. Abgene™ microtiter plates are manufactured to exacting specifications in Class 100000 cleanroom ISO 9001 conditions using high-quality medical grade virgin USP <661.1> polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handlers for high-throughput workflows. These polypropylene storage plates are offered with spindle, pyramidal, U, and V bottom wells along with a wide range of volumes to accommodate most applications.
Packaging: Abgene™ Storage plates are available individually or bulk packed.
Documents
Specifications
- No. of wells:96
- Sterility:Cleanroom-manufactured
- Color:Natural
- Description:Storage plate (Bulk)
- Well shape:Round
- Bottom style:V-conical
- Total Volume:800 µl



