Buffer Solutions
Buffer solution, 10.01
Catalog # 77679-414
Supplier: Ohaus
Supplier #: 30100429
Jump to:
Product Details & Documents
Buffer solution, 250 ml bottle.
Specifications
Specifications
- pH:10.01
- Density:Buffer solution
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



