Buffer Solutions
Buffer solution, 12.45
Catalog # 77679-416
Supplier: Ohaus
Supplier #: 30100440
Jump to:
Product Details & Documents
Buffer solution, 250 ml bottle.
Specifications
Specifications
- pH:12.45
- Density:Buffer solution
Buffer solution, 250 ml bottle.