Buffer Solutions
Buffer solution, 6.86
Catalog # 77679-408
Supplier: Ohaus
Supplier #: 30100426
Jump to:
Product Details & Documents
Buffer solution, 250 ml bottle.
Specifications
Specifications
- pH:6.86
- Density:Buffer solution
Buffer solution, 250 ml bottle.