Orders LinkContactUsLinkComponent
DissecTable Dissection Frames
Dissectable dissection board, large, 23×16×0.5"
Catalog # 77666-348
 :  WP-620
DissecTable Dissection Frames
Catalog # 77666-348
DissecTable Dissection Frames
Dissectable dissection board, large, 23×16×0.5"
Price
$434.52
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
DissecTable™ Dissection Board is a new and unique approach that makes this dissecting board more convenient than any other found on the market today. Made of heavy-duty stain resistant thick polyethylene, it will last for years to come without, bending.
  • Clean, safe, efficient way of handling infectious tissue specimens
  • Polyethylene
  • Spill grooves
  • Rubber feet
  • Heavy duty base for elevation
  • Stain resistant
  • Lasts for years
Double sided with one large area or two smaller areas on the opposite side. Dissection boards for use pathology labs, schools, forensic labs and more.
Specifications
  • Description:
    Dissectable dissection board, large
  • L×W×H:
    23×16×0.5"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.