Orders LinkContactUsLinkComponent
Sterilisation Baskets, Side Perforated, Flat Base | Avantor
Sterilisation Baskets, Side Perforated, Flat Base
Stainless steel 304, 540×250×60 mm
Catalog # 77666-174
Fournisseur #:  WP-4562
Sterilisation Baskets, Side Perforated, Flat Base
Catalog # 77666-174
Sterilisation Baskets, Side Perforated, Flat Base
Stainless steel 304, 540×250×60 mm
Price
$240.16
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Stainless steel sterilization baskets for instrument cleaning, sterilization and storage. The perforated side walls protect instrument edges and tips from damage. These sterilization baskets feature electropolished smooth, rounded corners. The baskets are burr free to protect wraps and prevent injury.
Specifications
  • Dimensions:
    540×250×60 mm
  • Material:
    Stainless steel 304
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.