Orders LinkContactUsLinkComponent
Reference Cell Electrodes
Reference Cell Electrodes
Reference cell with female connector
Catalog # 77665-106
Supplier #:  RC2F
Reference Cell Electrodes
Catalog # 77665-106
Reference Cell Electrodes
Reference cell with female connector
Price
$164.32
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
New, improved sintered pellets with lower resistance and high strength. Stable and well balanced in the presence of current, these small and inexpensive half-cells are easy to work with as bath electrodes.
  • Reference cell electrodes for laboratory research
Specifications
  • Description:
    Reference cell with female connector
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)