Orders LinkContactUsLinkComponent
Standard Rectangular Quartz Cuvettes
Standard Rectangular Quartz Cuvettes
Standard rectangular cuvette, 4.5×12.5×45 mm, beam width 10 mm, 0.7 ml, Quartz
Catalog # 77664-180
Supplier #:  CUV2102-1
Standard Rectangular Quartz Cuvettes
Catalog # 77664-180
Standard Rectangular Quartz Cuvettes
Standard rectangular cuvette, 4.5×12.5×45 mm, beam width 10 mm, 0.7 ml, Quartz
Price
$357.06
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Glass and synthetic quartz cuvettes are ideal for UV/VIS/NIR absorbance or fluorescence experiments. Synthetic quartz can be used in deep UV applications and is recommended for fluorescent applications, as it does not exhibit background fluorescence. Quartz cuvettes (absorbance, fluorescence and flow) are shipped individually packaged. These economic quartz are ideal for precision measurements because of their high quality materials used and their low manufacturing tolerances.
Spectral Range (>80%): 200 to 2500 nm; Transmission Difference between different cuvettes: Less than 1% (Typical transmission curves of synthetic quartz cuvettes were measured empty, thickness 1.25 mm × 2, including surface reflections, measured with a TIDAS II against air as reference).
Specifications
  • Volume:
    0.7 ml
  • Description:
    Standard rectangular cuvette, 4.5×12.5×45 mm, beam width 10 mm
  • Material:
    Quartz
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)