Orders LinkContactUsLinkComponent
Microelectrode Holders
Microelectrode Holders
1.2 mm, Microelectrode holder (MEH3SFW)
Catalog # 77664-722
Supplier #:  MEH3SFW12
Microelectrode Holders
Catalog # 77664-722
Microelectrode Holders
1.2 mm, Microelectrode holder (MEH3SFW)
Price
$142.22
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
WPI's microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers. A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets.
  • Female connector
  • Wire half-cell
  • No pressure port
  • Screw cap
The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps). Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂. Holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters. (Call WPI regarding custom designs for other glass diameters.) The holder style you select will depend on your experimental application, space, and instrumentation.
Specifications
  • Description:
    Microelectrode holder (MEH3SFW)
  • Length:
    1.2 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)