Sterilization Mesh Cassettes
Product Details & Documents
Mesh sterilization baskets are perfect for sterilizing, storing and organizing small components or instrument. These baskets are manufactured from 304 stainlesss steel. They are non-magnetic and electropolished.
- These baskets come with a wire locking clip
- This style of sterilization baskets is available in three sizes.
They are burr-free to protect the instrument wraps from tearing. The fine mesh has open squares of 1.6 mm, and the wire edge diameter is 0.7 mm.
Specifications
- Dimensions:8×8×3.4 cm (3.1×3.1×1.4")
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)