Iris Forceps, Disposable
ris Forceps are a pair of surgical forceps (tweezers) often used in ophthalmic procedures. Iris Forceps are a small pair of forceps with a fine pointed tip for grasping delicate tissue with minimal trauma.
- Narrow, serrated jaws of the surgical forceps
- Fine tipped tweezers
- Surgical grade stainless steel
The narrow, serrated jaws of the surgical forceps allow for fine manipulation of structures like the iris of the eye. Researchers like these fine tipped tweezers for precision work in microdissection. These forceps are 10 cm long with fine tips, and they come with straight or curved tips. The Iris Forceps are manufactures with surgical grade stainless steel.
Our Disposable Iris Forceps are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. These economy forceps are sold in packages of 12, perfect for students or for disposable use. All sales are final on disposable instrument packs.
- Description:Iris forcep, serrated
- Length:10 cm (4")
- Material:Stainless steel
- Curvature:Curved
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.