Orders LinkContactUsLinkComponent
Filling Solutions
Filling Solutions
Filling solution, 10 ml, Nitric oxide
Catalog # 77663-932
Supplier #:  7325
Filling Solutions
Catalog # 77663-932
CAS Number:  
Filling Solutions
Filling solution, 10 ml, Nitric oxide
CAS Number:  
Price
$160.61
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Filling solutions to use with ISO-NOP, ISO-OXY-2 or OXELP oxygen sensors.
Specifications
Specifications
  • Description:
    Filling solution
  • Size:
    10 ml
  • Fill solution:
    Nitric oxide
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.