Orders LinkContactUsLinkComponent
Derf Needle Holders
Derf needle holder, German, 12 cm (5")
Catalog # 77662-492
 :  501246-G
Derf Needle Holders
Catalog # 77662-492
Derf Needle Holders
Derf needle holder, German, 12 cm (5")
Price
$101.10
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Derf Needle Holders are surgical instruments designed for holding needles during suturing.
  • Short serrated jaws
  • Interlocking ratchet handles
  • Surgical grade stainles steel or premium German steel
  • Straight
The small size 12.5 cm (5") and the short jaws are perfect for smaller needles like 5-0 or 6-0 sutures. The tip has fine serrations and a groove in the center. The groove helps to securely hold thicker surgical needle. The tips of needle holders are generally shorter than those of surgical forceps. The interlocking ratchet handle ensures a firm grasp of the needle when the lock is engaged.
The jaws are straight. These needle holders are manufactured from surgical grade stainless steel or premium German steel, both of which are autoclavable or suitable for chemical sterilization. Always clean your surgical instrument thoroughly every time you use them.
Specifications
  • Description:
    Derf needle holder, German
  • Length:
    12 cm (5")
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.