Orders LinkContactUsLinkComponent
Absorbent Paper Points
Absorbent Paper Points
White, Extra fine, Pack of 200
Catalog # 77663-542
Supplier #:  504180
Absorbent Paper Points
Catalog # 77663-542
Absorbent Paper Points
White, Extra fine, Pack of 200
Price
$63.28
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Absorbent paper points are commonly used in laboratories, particularly in the field of dentistry and endodontics. These paper points are small, sterile, and highly absorbent.
  • Made from higly absorbent paper
  • Uniformly rolled to ensure correct size
  • Good for small volume absorption in confined areas
These sterile paper points are tapered to target specific areas and soak up small volumes of liquid. A variety of color-coded sizes are available, including extra fine, #15 fine, medium and coarse.
Packaging: Package of 100 to 200, depending on the size of the paper points.
Specifications
  • Color:
    White
  • Packaging:
    Pack of 200
  • Type:
    Extra fine
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.