Orders LinkContactUsLinkComponent
Forceps, Allis Tissue
Forceps, Allis Tissue
Forceps, Allis tissue, german, 6"
Catalog # 77662-594
Supplier #:  501324-G
Forceps, Allis Tissue
Catalog # 77662-594
Forceps, Allis Tissue
Forceps, Allis tissue, german, 6"
Price
$151.66
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
German made instruments are known for quality, because of their precision manufacturing and attention to details.
  • 15 cm (6") Long
  • 4×5 teeth
  • Stainless steel
Specifications
  • Description:
    Forceps, Allis tissue, german
  • Length:
    6"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)