VWR® Ion Selectrive Electrodes
VWR® ISE05 ion selective electrode, combination, BNC (Q9), Plastic, 1.0×10⁻⁷ - 0.1 M, 0…80 °C, Cupric (Cu²⁺)
Catalog # 77618-840
Supplier: VWR International
Jump to:
Product Details & Documents
The VWR® Ion Selective Electrodes (ISE) allow for the measurement of concentration of a specific ion.
- ISE probe and reference electrode combined
- BNC connection
Includes ISE reference filler solution.
Specifications
- Temperature range:0…80 °C
- Description:VWR® ISE05 ion selective electrode, combination
- Measurement range:1.0×10⁻⁷ - 0.1 M
- Body material:Plastic
- Ion:Cupric (Cu²⁺)
- Connector:BNC (Q9)