KIT MHC TETRAMER HLA-AX03:01 150 TESTS
Catalog # MSPP-101601150
Supplier: Eagle Biosciences
KIT MHC TETRAMER HLA-AX03:01 150 TESTS
Catalog # MSPP-101601150
Supplier: Eagle Biosciences
Supplier Number: 1016-01-150
Frequently Bought Together
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Dichloromethane ≥99.9% stabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Methanol ≥99.8% ACS for HPLC, Supelco®
Supplier: MilliporeSigma
Tetrahydrofuran ≥99.9% (by GC) unstabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Masterflex® I/P® High-Performance Precision Pump Tubing, Versilon™ A-60-G, Avantor®
Supplier: VWR International
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
1-Decanol 98+%
Supplier: Thermo Scientific Chemicals
Cryoscope Standards, Advanced Instruments
Supplier: Advanced Instruments
Calcium Nitrate 4-Hydrate
Supplier: Ward's Science
1-Pentanesulfonic acid sodium salt monohydrate for HPLC
Supplier: Thermo Scientific Chemicals
Human Recombinant PDGF-AA (from <i>E. coli</i>)
Supplier: Peprotech
Mercury(II) bromide ACS
Supplier: Thermo Scientific Chemicals
Q5® Site-Directed Mutagenesis Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
Single-Use Needles, BD Medical
Supplier: BD
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA