Order Entry
Canada
Orders LinkContactUsLinkComponent
 

LabExact® Extraction Thimbles, Cellulose

Cotton linter cellulose, LabExact® Extraction thimbles cellulose, with lid, 30×80 mm, 1.5 mm, 5 µm
  77590-876
 :  LECT5220
LabExact® Extraction Thimbles, Cellulose
  77590-876
LabExact® Extraction Thimbles, Cellulose
LabExact® Extraction Thimbles, Cellulose

Cotton linter cellulose, LabExact® Extraction thimbles cellulose, with lid, 30×80 mm, 1.5 mm, 5 µm

 
$310.97
   
Box of 25
 

LabExact® high quality cellulose extraction thimbles are available in a variety of dimensions to fit most Soxhlet extraction units.

  • Extraction thimbles are manufactured with high purity cellulose fibers and have a consistent wall thickness
  • Available as fast flow, with lid and extra density

Common applications include air and waste gas analysis, collection of solid particles such as dust, concrete slurry evaluation, analysis of pesticide residues, oil/fat content of solid foods and oil and grease analysis of solid wastes.

 
  • Wall thickness:
    1.5 mm
  • Description:
    LabExact® Extraction thimbles cellulose, with lid
  • Material:
    Cotton linter cellulose
  • I.D.×H:
    30×80 mm
  • Particle retention:
    5 µm