LabExact® LabExact® Bench Protector Papers
Catalog # 77590-484
Supplier: I.W. TREMONT CO., INC.
Supplier #: LEBPS4657
Product Details & Documents
LabExact® disposable, highly absorbent pure cellulose paper with polyethylene backing protects laboratory surfaces against liquid spills and leakage through to the bench top.
- Lays flat with less wrinkles for sturdy glassware use
- Tear resistant material, whether wet or dry
- The polyethylene backing material is skid resistant, waterproof and chemical resistant
- The absorbent paper quickly traps spilled or drained fluids
- Smooth white surface is easy to write on
It is designed to protect laboratory surfaces against liquid spills and prevent leakage through to the bench top, necessary to maintain a safer work environment.
Certifications: Meets OSHA regulation 29CFR 1910.1030 for occupational exposure to bloodborne pathogens.
Documents
TDS Bench Protector Paper.pdf