Orders LinkContactUsLinkComponent
PathFlow PiSmart Laser Cassettes, Threaded
PathFlow PiSmart Laser Cassettes, Threaded
PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded, Green
Catalog # 77575-548
Supplier:   StatLab
Supplier #:  CAS-LPI100BX-GR
PathFlow PiSmart Laser Cassettes, Threaded
Catalog # 77575-548
Supplier:  StatLab
PathFlow PiSmart Laser Cassettes, Threaded
PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded, Green
Price
$1,419.00
Log in for availability
Case of 2,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pathflow PiSmart laser cassettes are a two piece design with a rear hinge and front tab closure. Threaded cassettes come in stacks of 40 cassettes, and 50 stacks per case of 2000.
These threaded cassettes can be easily and quickly loaded in the PiSmart Laser Printers. These cassettes are a validated consumable for the PiSmart laser cassette printer.
Specifications
  • Color:
    Green
  • Description:
    PathFlow PiSmart laser, separate lid with rear hinge and front closure, threaded
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)