Orders LinkContactUsLinkComponent
Cleanroom Labels, Core2Write
Cleanroom Labels, Core2Write
Core2Write label, sterile, irradiated, 76,2×25,4 mm, 18 labels/sheet, 500 sheets/case
Catalog # 76627-991
Supplier:   Veltek Associates
Supplier #:  C2WR-3X1-SHEET-02
Cleanroom Labels, Core2Write
Catalog # 76627-991
Supplier:  Veltek Associates
Cleanroom Labels, Core2Write
Core2Write label, sterile, irradiated, 76,2×25,4 mm, 18 labels/sheet, 500 sheets/case
Price
$5,480.00
Log in for availability
Case of 5,000
Product Details & Documents
Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures. All labels can be customised or left blank.
Specifications
  • Description:
    Core2Write label, sterile, irradiated
  • Dimensions:
    76,2×25,4 mm
  • Packaging:
    18 labels/sheet, 500 sheets/case